Please login to your online account to display your discounted pricing

Alfa Aesar™ Bone Morphogenetic Protein 4, Human, Inactive Protein

For research use

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements -30° to -10°C
Source Escherichia coli
Product Type Bone Morphogenetic Protein 4, Human, inactive protein
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67416AME

 
alfa aesar™
J67416AME
2μg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67416EXE

 
alfa aesar™
J67416EXE
10μg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements -30° to -10°C
Source Escherichia coli
Product Type Bone Morphogenetic Protein 4, Human, inactive protein
Format Lyophilized powder
Regulatory Status RUO