Please login to your online account to display your discounted pricing

Alfa Aesar™ Bone Morphogenetic Protein 2, Human


For research use

Manufacturer: alfa aesar™  J67332AME

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67332AME

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Format Lyophilized powder
Recombinant No
Source Escherichia coli
Product Type Bone Morphogenetic Protein 2, Human
Regulatory Status RUO
Quantity 2μg
Formulation Lyophilized powder