Please login to your online account to display your discounted pricing

Alfa Aesar™ Bone Gla Protein (45-49)

For cellular and molecular biology applications

Specifications

Product Type Bone Gla Protein (45-49)
Molecular Weight 581.66
Storage Requirements -30° to -10°C
Sequence Phe-Tyr-Gly-Pro-Val
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66249LB0

 
alfa aesar™
J66249LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66249MCR

 
alfa aesar™
J66249MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Bone Gla Protein (45-49)
Molecular Weight 581.66
Storage Requirements -30° to -10°C
Sequence Phe-Tyr-Gly-Pro-Val
Format Solid
Common Name Bone γ-Carboxyglutamate Protein Fragment (45-49)
Regulatory Status RUO
Purification Method HPLC