missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Bolt™ 4 to 12%, Bis-Tris, 1.0 mm Mini Protein Gel, 17-well DFS Item

Separation Range: 3.5 to 260 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini

Manufacturer:  Invitrogen™ LSNW04127BOX

Catalog No. NW04127BOX



Description

Description

Similar to NuPAGE™ Bis-Tris gels, Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for Western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt Bis-Tris Plus gels to obtain optimal results for day-to-day protein separation needs.
Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and 4–12% gradient. The gels can be run using Bolt MES or MOPS SDS running buffer to obtain different separation ranges. Bolt Bis-Tris Plus gels consist of a neutral pH-buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0. Bolt Bis-Tris Plus gels also come in four different well formats: 10, 12, 15, and 17 wells.

  • Optimized run conditions—separate proteins using constant voltage in approximately 35 minutes
  • Better protein integrity—neutral pH sample preparation process preserves proteins
  • Improved protein separation—offer sharper bands, high resolution, longer shelf life, and higher accuracy of results than tris-glycine—based gels
  • Wedge-shaped wells—easier sample loading and up to two times higher sample volume than other 1.0mm-thick gels

  • Multichannel pipettor compatible—achieve high throughput sample preparation and loading using the 17-well format

Specifications

Specifications

Day-to-Day Protein Separations, Techniques where Protein Integrity is Crucial, Western Blot Transfer and Analysis
8 cm
4°C to 25°C
8 cm
Bis-Tris Plus Gel
3.5 to 260 kd
Bis-Tris
1.0 mm
Protein
1 Box
Up to 30 µL per well
Room Temperature
1.0 mm
Plastic
16 Months
up to 30 µL
4-12% acrylamide
8 cm
Up to 30 μL/well (15 μL recommended)
8 cm
Mini
4 to 12%
Denaturing
Mini Gel Tank,XCell SureLock Mini
17-well
No Adapters
Molecular Weight
1D well (wedge shaped)
Bolt™
Gradient Page
Denaturing
SDS
Documents
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.