missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boekel Scientific™ Ultem™ 1000 Pipet Cans

Ideal for sterilizing and containing pipets used in work sensitive to metallic ion contaminants

Supplier:  Boekel Scientific™ 135909

Catalog No. 03-465-2



Description

Description

  • Patented pipet cans are fabricated from Ultem 1000 polyetherimide resin that withstands repeated steam and dry-heat sterilizations
  • Transparent for easy visibility of contents; easy to clean
  • Flat edge on base prevents rolling

Order Info

Choice of three heights; all have a 2 in. diameter (5cm).

Specifications

Specifications

9 in.
2 in.
Polyetherimide Resin
Round
22.9 cm
5.1cm
For sterilizing and storing pipets used in procedures sensitive to Metallic ion contamination
Pipet Can
SDS
Documents

Documents

Promotions

Promotions