Boekel Scientific TropiCooler™ Incubator Blocks
Ideal for restriction enzymes and DNA libraries
$187.70 - $241.62
Specifications
Material Aluminum
Product Type Block module with block extractor
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
05-450-3 Boekel Scientific
BKL260052
Boekel TropiCooler incubator Each for $187.70
05-450-4 Boekel Scientific
BKL260152
Boekel TropiCooler incubator Each for $193.15
13-245-114 Boekel Scientific
BKL260055
Boekel TropiCooler incubator Each for $241.62
Specifications
Description
  • Machined aluminum blocks with anodized finish and self-contained recessed handle
  • Hold 0.5 and 1.5mL microcentrifuge tubes (13-245-114 holds seven of each tube size) Dimensions, 21/8L × 31/4W × 11/2 in.H (5.4 × 8.3 × 3.8cm)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Aluminum
Block module with block extractor