Boekel Scientific™ TropiCooler™ Incubator Blocks
Ideal for restriction enzymes and DNA libraries
Manufacturer:  Boekel Scientific™ BKL260152
 View more versions of this product
Catalog No. 05-450-4
Description
Description
  • Machined aluminum blocks with anodized finish and self-contained recessed handle
  • Hold 0.5 and 1.5mL microcentrifuge tubes (13-245-114 holds seven of each tube size) Dimensions, 21/8L × 31/4W × 11/2 in.H (5.4 × 8.3 × 3.8cm)
Specifications
Specifications
Boekel TropiCooler incubator
Aluminum
For 1.5mL microcentrifuge tube
Block module with block extractor
Documents