Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with Temperature Probe
A flexible device for measuring Temperature
Manufacturer:  Boekel Scientific™ BKL555000
Catalog No. 11-700-150
Includes: Open Source Transmitter and Temperature probe
Description
Description
  • Built around UNO, an open source circuit board that allows easy user programming and experiment control
  • Repairable and industrial design supports years of experiments
  • External I/O connector for connection to pumps or alarms
  • User friendly probe calibration menus
  • USB drive data logging
  • Large color screen
  • Also works with pH, DO and EC probes
Warranty
Two years.
Specifications
Specifications
Open Source Transmitter and Temperature probe
OSB Transmitter with Temperature probe
2 Years
Large Screen