missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boekel Scientific™ Open Source Bio™ (OSB) Transmitter: Calibration Reagents

For use with Open Source Bio (OSB) Transmitter.

Manufacturer:  Boekel Scientific™ BKL9510020

 View more versions of this product

Catalog No. 11-700-157



Description

Description

For use with Open Source Bio (OSB) Transmitter.
Specifications

Specifications

pH Calibration Reagents
Open Source Bio (OSB) Transmitter
SDS
Documents
Product Certifications
Promotions

Promotions