Please login to your online account to display your discounted pricing

BOC-trans-4-hydroxy-L-proline, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC435480050

 View more versions of this product

Catalog No. AC435480050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.5%
Formula Weight 231.25
Quantity 5g
Color White to off-white
Melting Point 121° to 125°C
Specific Rotation -51.5° to -55.5° (20°C, 589nm) (c=1 in DMF)
Chemical Name or Material BOC-trans-4-Hydroxy-L-proline
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H17NO5
CAS 13726-69-7
Synonym trans-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline
Elemental Analysis 50.6 to 53.2% C, 5.9 to 6.2% N
Infrared Spectrum Authentic

Specifications

Chemical Name or Material BOC-trans-4-Hydroxy-L-proline
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H17NO5
Formula Weight 231.25
Synonym trans-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline