missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-Sarcosine, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 275670250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
BOC-Sarcosine | |
13734-36-6 | |
99% min. (ELSD) (HPLC) | |
C8H15NO4 | |
MFCD00037795 | |
boc-sar-oh, boc-sarcosine, n-boc-sarcosine, glycine, n-1,1-dimethylethoxy carbonyl-n-methyl, n-tert-butoxycarbonyl-n-methylglycine, t-boc-sarcosine, n-t-boc-sarcosine, 2-tert-butoxy carbonyl methyl amino acetic acid, n-tert-butoxycarbonylsarcosine, tert-butoxycarbonyl methyl amino acetic acid | |
YRXIMPFOTQVOHG-UHFFFAOYSA-N | |
2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid | |
83692 | |
99+% |
99+% | |
Authentic | |
Glass bottle | |
(CH3)3COCON(CH3)CH2COOH | |
25g | |
Solubility in water: slightly soluble | |
CC(C)(C)OC(=O)N(C)CC(=O)O | |
189.21 | |
189.21 |
Chemical Identifiers
13734-36-6 | |
189.21 | |
YRXIMPFOTQVOHG-UHFFFAOYSA-N | |
83692 | |
CC(C)(C)OC(=O)N(C)CC(=O)O |
C8H15NO4 | |
MFCD00037795 | |
boc-sar-oh, boc-sarcosine, n-boc-sarcosine, glycine, n-1,1-dimethylethoxy carbonyl-n-methyl, n-tert-butoxycarbonyl-n-methylglycine, t-boc-sarcosine, n-t-boc-sarcosine, 2-tert-butoxy carbonyl methyl amino acetic acid, n-tert-butoxycarbonylsarcosine, tert-butoxycarbonyl methyl amino acetic acid | |
2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 237-306-0
TSCA : TSCA
RUO â Research Use Only