Learn More
BOC-ON, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 185890250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
BOC-ON | |
58632-95-4 | |
White to Beige | |
Authentic | |
Glass bottle | |
(CH3)3COCO2N=C(C6H5)CN | |
25 g | |
15, 1323 | |
Solubility in water: insoluble. Other solubilities: soluble in ethyl acetate,ether,benzene,dioxane,,chloroform,acetone,methanol,isopropanol and,tert-butanol,insoluble in petroleum ether | |
CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1 | |
246.27 | |
246.27 | |
Fine Crystalline Powder |
99% | |
86°C to 89°C | |
>100°C | |
99% | |
C13H14N2O3 | |
MFCD00001863 | |
06,91; 10,61 | |
2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino | |
QQWYQAQQADNEIC-PTNGSMBKSA-N | |
tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate | |
5868400 | |
99% |
Chemical Identifiers
58632-95-4 | |
246.27 | |
QQWYQAQQADNEIC-PTNGSMBKSA-N | |
5868400 | |
CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1 |
C13H14N2O3 | |
MFCD00001863 | |
2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino | |
tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapo
GHS Signal Word: Warning
EINECSNumber : 261-370-9
TSCA : TSCA
RUO – Research Use Only