missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-O-Benzyl-L-threonine hydroxysuccinimide ester, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440900010

 View more versions of this product

Catalog No. AC440900010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

BOC-O-Benzyl-L-threonine hydroxysuccinimide ester
Glass Bottle
1% Max.
104°C to 107°C
32886-43-4
C20H26N2O7
SNEHQGZSMYSVFT-DYVFJYSZSA-N
(2,5-dioxopyrrolidin-1-yl) (2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoate
12879029
Powder
Authentic
10.6
White
1g
97.5% min. (HPLC)
2,5-dioxoazolidinyl 2s,3r-2-tert-butoxy carbonylamino-3-phenylmethoxy bu tanoate, 2,5-dioxopyrrolidin-1-yl 2s,3r-3-benzyloxy-2-tert-butoxycarbonyl amino butanoate, 2s,3r-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino butanoate, boc-o-benzyl-l-threonine hydroxysuccinimide ester, boc-thr bzl-osu, boc-threonine bzl-osu, n-alpha-t-butyloxycarbonyl-o-benzyl-l-threonine succinimidyl ester, n-tert-butoxycarbonyl-o-benzyl-l-threonine succinimidyl ester
CC(C(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C)OCC2=CC=CC=C2
406.44
406.44
98%

Chemical Identifiers

32886-43-4
406.44
2,5-dioxoazolidinyl 2s,3r-2-tert-butoxy carbonylamino-3-phenylmethoxy bu tanoate, 2,5-dioxopyrrolidin-1-yl 2s,3r-3-benzyloxy-2-tert-butoxycarbonyl amino butanoate, 2s,3r-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino butanoate, boc-o-benzyl-l-threonine hydroxysuccinimide ester, boc-thr bzl-osu, boc-threonine bzl-osu, n-alpha-t-butyloxycarbonyl-o-benzyl-l-threonine succinimidyl ester, n-tert-butoxycarbonyl-o-benzyl-l-threonine succinimidyl ester
(2,5-dioxopyrrolidin-1-yl) (2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoate
C20H26N2O7
SNEHQGZSMYSVFT-DYVFJYSZSA-N
12879029
CC(C(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C)OCC2=CC=CC=C2
SDS Documents