Please login to your online account to display your discounted pricing

BOC-O-Benzyl-L-threonine, 99%, Acros Organics

Chemical Identifiers

Chemical Name or Material BOC-O-Benzyl-L-threonine
Assay Percent Range 99%
Molecular Formula C16H23NO5
Formula Weight 309.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC442000010

 
thermo fisher scientific
AC442000010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442000050

 
thermo fisher scientific
AC442000050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.37
Quantity 1g
CAS 15260-10-3
Molecular Formula C16H23NO5
Assay Percent Range 99%
Chemical Name or Material BOC-O-Benzyl-L-threonine