missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-O-Benzyl-L-threonine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442000010

 View more versions of this product

Catalog No. AC442000010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

BOC-O-Benzyl-L-threonine
15.5
1g
98.5% min. (HPLC)
2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid, boc-o-benzyl-l-threonine, boc-thr bzl, boc-thr bzl-oh, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, n-boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, o-benzyl-n-tert-butoxycarbonyl-l-threonine
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
309.37
309.37
Glass Bottle
115°C to 117°C
15260-10-3
C16H23NO5
CTXPLTPDOISPTE-YPMHNXCESA-N
(2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
1549483
99%

Chemical Identifiers

15260-10-3
309.37
2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid, boc-o-benzyl-l-threonine, boc-thr bzl, boc-thr bzl-oh, l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine, n-boc-o-benzyl-l-threonine, n-tert-butoxycarbonyl-o-benzyl-l-threonine, o-benzyl-n-tert-butoxycarbonyl-l-threonine
(2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
C16H23NO5
CTXPLTPDOISPTE-YPMHNXCESA-N
1549483
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
Safety and Handling

Safety and Handling

EINECSNumber : 239-304-5

SDS Documents