missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-O-Benzyl-L-serine hydroxysuccinimide ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440890050

 View more versions of this product

Catalog No. AC440890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

BOC-O-Benzyl-L-serine hydroxysuccinimide ester
Glass bottle
1% max.
107°C to 109°C
13650-73-2
C19H24N2O7
NTFYOALQRQBBDG-AWEZNQCLSA-N
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoate
12879025
Powder
Authentic
-16.4
White
5g
96% min. (HPLC)
2,5-dioxopyrrolidin-1-yl 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, 2,5-dioxopyrrolidin-1-yl o-benzyl-n-t-butoxycarbonyl-l-serinate, boc-ser bzl osu, boc-ser bzl-osu, boc-ser bzl-osu hplc, n-boc-3-benzyloxy-ala-succinimidyl oh, n-boc-o-benzyl-l-serine n-succinimidyl ester, n-tert-butoxycarbonyl-o-benzyl-l-serine n-succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino propanoate
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
392.41
392.41
97%

Chemical Identifiers

13650-73-2
392.41
2,5-dioxopyrrolidin-1-yl 2s-3-benzyloxy-2-tert-butoxycarbonyl amino propanoate, 2,5-dioxopyrrolidin-1-yl o-benzyl-n-t-butoxycarbonyl-l-serinate, boc-ser bzl osu, boc-ser bzl-osu, boc-ser bzl-osu hplc, n-boc-3-benzyloxy-ala-succinimidyl oh, n-boc-o-benzyl-l-serine n-succinimidyl ester, n-tert-butoxycarbonyl-o-benzyl-l-serine n-succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 3-benzyloxy-2-tert-butoxycarbonyl amino propanoate
(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoate
C19H24N2O7
NTFYOALQRQBBDG-AWEZNQCLSA-N
12879025
CC(C)(C)OC(=O)NC(COCC1=CC=CC=C1)C(=O)ON2C(=O)CCC2=O
Documents