Please login to your online account to display your discounted pricing

BOC-O-Benzyl-L-serine hydroxysuccinimide ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material BOC-O-Benzyl-L-serine hydroxysuccinimide ester
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C19H24N2O7
Formula Weight 392.41
Synonym BOC-Ser(Bzl)-OSu
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC440890050

 
thermo fisher scientific
AC440890050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440890250

 
thermo fisher scientific
AC440890250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 392.41
Quantity 5g
Color White to off-white
Water 1% max.
Chemical Name or Material BOC-O-Benzyl-L-serine hydroxysuccinimide ester
Packaging Glass bottle
Specific Rotation -14° to -18° (20°C, 589nm) (c=1 in DMF)
Assay Percent Range 97%
Molecular Formula C19H24N2O7
CAS 13650-73-2
Infrared Spectrum Authentic
Synonym BOC-Ser(Bzl)-OSu