Please login to your online account to display your discounted pricing

BOC-O-Benzyl-L-serine hydroxysuccinimide ester, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC440890250

 View more versions of this product

Catalog No. AC440890250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 392.41
Assay ≥96.0%
Physical Form Powder
Quantity 25g
Color White to off-white
Specific Rotation -14° to -18° (20°C, 589nm) (c=1 in DMF)
Water 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
CAS 13650-73-2
Molecular Formula C19H24N2O7
Chemical Name or Material BOC-O-Benzyl-L-serine hydroxysuccinimide ester
Packaging Glass bottle
Synonym BOC-Ser(Bzl)-OSu

Specifications

Chemical Name or Material BOC-O-Benzyl-L-serine hydroxysuccinimide ester
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C19H24N2O7
Formula Weight 392.41
Synonym BOC-Ser(Bzl)-OSu