Please login to your online account to display your discounted pricing

BOC-N-methyl-D-alanine, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC434240010

 View more versions of this product

Catalog No. AC434240010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 203.24
Quantity 1g
Color White
Melting Point 91° to 95°C
Chemical Name or Material BOC-N-Methyl-D-alanine
Assay Percent Range 97%
Specific Rotation 29° to 31° (20°C, 589nm) (c=1, ethanol)
Molecular Formula C9H17NO4
MDL Number MFCD00063137
CAS 19914-38-6
Infrared Spectrum Authentic

Specifications

Chemical Name or Material BOC-N-Methyl-D-alanine
CAS 19914-38-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H17NO4
Formula Weight 203.24
MDL Number MFCD00063137