Please login to your online account to display your discounted pricing

BOC-N-methyl-D-alanine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material BOC-N-Methyl-D-alanine
CAS 19914-38-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H17NO4
Formula Weight 203.24
MDL Number MFCD00063137
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC434240010

  SDS
thermo fisher scientific
AC434240010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434240050

  SDS
thermo fisher scientific
AC434240050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 203.24
Assay ≥96.0%
Quantity 1g
Color White
Melting Point 91° to 95°C
Assay Percent Range 97%
Specific Rotation 29° to 31° (20°C, 589nm) (c=1, ethanol)
Chemical Name or Material BOC-N-Methyl-D-alanine
Molecular Formula C9H17NO4
MDL Number MFCD00063137
CAS 19914-38-6
Infrared Spectrum Authentic