Please login to your online account to display your discounted pricing

BOC-α-Methylalanine, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338790250

 View more versions of this product

Catalog No. AC338790250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 117° to 121°C
Color White
Quantity 25g
Chemical Name or Material BOC-α-Methylalanine
CAS 30992-29-1
Physical Form White crystalline powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C9H17NO4
Linear Formula (CH3)3COCONHC(CH3)2COOH
Formula Weight 203.24g/mol
MDL Number MFCD00042973

Specifications

Chemical Name or Material BOC-α-Methylalanine
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C9H17NO4
Linear Formula (CH3)3COCONHC(CH3)2COOH
Formula Weight 203.24g/mol
MDL Number MFCD00042973