Please login to your online account to display your discounted pricing

BOC-α-Methylalanine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material BOC-α-Methylalanine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H17NO4
Linear Formula (CH3)3COCONHC(CH3)2COOH
Formula Weight 203.24
MDL Number MFCD00042973
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC338790010

  SDS
fisher scientific acros
338790010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 203.24
Quantity 1g
Color White
Melting Point 117° to 121°C
Packaging Glass bottle
Molecular Formula C9H17NO4
Linear Formula (CH3)3COCONHC(CH3)2COOH
Chemical Name or Material BOC-α-Methylalanine
Assay Percent Range 98%
MDL Number MFCD00042973
CAS 30992-29-1
Infrared Spectrum Authentic