Please login to your online account to display your discounted pricing

BOC-α-Methylalanine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  338790010

 View more versions of this product

Catalog No. AC338790010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 203.24
Quantity 1g
Color White
Melting Point 117° to 121°C
Linear Formula (CH3)3COCONHC(CH3)2COOH
Packaging Glass bottle
Molecular Formula C9H17NO4
CAS 30992-29-1
Assay Percent Range 98%
Chemical Name or Material BOC-α-Methylalanine
MDL Number MFCD00042973
Infrared Spectrum Authentic

Specifications

Chemical Name or Material BOC-α-Methylalanine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H17NO4
Linear Formula (CH3)3COCONHC(CH3)2COOH
Formula Weight 203.24
MDL Number MFCD00042973