missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Tyrosine, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC275701000
Specifications
BOC-L-Tyrosine | |
99+% | |
HOC6H4CH2CH(COOH)NHCO2C(CH3)3 | |
MFCD00037179 | |
Glass bottle | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O | |
281.308 | |
281.3 | |
Powder | |
1% Max. (K.F.) | |
White | |
100g |
3978-80-1 | |
C14H19NO5 | |
Authentic | |
boc-l-tyrosine, boc-tyr-oh, chembl65386, l-tyrosine, n-1,1-dimethylethoxy carbonyl, n-boc-l-tyrosine, n-boc-tyrosine, n-t-boc-l-tyrosine, n-t-butoxycarbonyl-l-tyrosine, n-tert-butoxycarbonyl tyrosine, n-tert-butoxycarbonyl-l-tyrosine | |
CNBUSIJNWNXLQQ-NSHDSACASA-N | |
(2S)-3-(4-hydroxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
117439 | |
+ 2.50 | |
≥99% | |
+2.5° (20°C c=2,acetic acid) | |
135.0°C to 140.0°C |
Chemical Identifiers
3978-80-1 | |
281.308 | |
CNBUSIJNWNXLQQ-NSHDSACASA-N | |
117439 | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O |
C14H19NO5 | |
MFCD00037179 | |
boc-l-tyrosine, boc-tyr-oh, chembl65386, l-tyrosine, n-1,1-dimethylethoxy carbonyl, n-boc-l-tyrosine, n-boc-tyrosine, n-t-boc-l-tyrosine, n-t-butoxycarbonyl-l-tyrosine, n-tert-butoxycarbonyl tyrosine, n-tert-butoxycarbonyl-l-tyrosine | |
(2S)-3-(4-hydroxyphenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |