missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-L-Serine methyl ester, 95%, ACROS Organics™

Chemical Identifiers

CAS 2766-43-0
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.24
InChI Key SANNKFASHWONFD-LURJTMIESA-N
Synonym boc-l-ser-ome, boc-l-serine methyl ester, boc-ser-ome, boc-ser-ome; boc-l-serine methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl tert-butoxycarbonyl-l-serinate, n-boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester
PubChem CID 7019177
IUPAC Name methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC437820010
SDS View Documents
Thermo Fisher Scientific
AC437820010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437820100
SDS View Documents
Thermo Fisher Scientific
AC437820100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2766-43-0
219.24
boc-l-ser-ome, boc-l-serine methyl ester, boc-ser-ome, boc-ser-ome; boc-l-serine methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl tert-butoxycarbonyl-l-serinate, n-boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester
methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
C9H17NO5
SANNKFASHWONFD-LURJTMIESA-N
7019177
CC(C)(C)OC(=O)NC(CO)C(=O)OC

Specifications

1.1000g/mL
Authentic
1.1
Colorless to Yellow
2766-43-0
C9H17NO5
SANNKFASHWONFD-LURJTMIESA-N
methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
7019177
Liquid or Oil
BOC-L-Serine methyl ester
113°C
Glass bottle
-18
1g
94% min. (GC)
boc-l-ser-ome, boc-l-serine methyl ester, boc-ser-ome, boc-ser-ome; boc-l-serine methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, methyl tert-butoxycarbonyl-l-serinate, n-boc-l-serine methyl ester, n-tert-butoxycarbonyl-l-serine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, serine, n-1,1-dimethylethoxy carbonyl-, methyl ester
CC(C)(C)OC(=O)NC(CO)C(=O)OC
219.24
219.24
95%
Safety and Handling

Safety and Handling

GHS H Statement:

GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS