missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Serine hydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 275660250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
84.0°C to 89.0°C | |
3262-72-4 | |
98% | |
Authentic | |
MFCD00037243 | |
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser | |
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O | |
− 8.10 | |
98766 | |
98% | |
7.5 to 8.0% (K.F.) | |
186°C |
BOC-L-Serine hydrate | |
White | |
C8H15NO5 | |
HOCH2CH(COOH)NHCOOC(CH3)3· | |
Glass Bottle | |
FHOAKXBXYSJBGX-MLHKIVSYNA-N | |
(2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
205.21 | |
205.21 | |
Technical | |
25 g | |
Powder |
Chemical Identifiers
3262-72-4 | |
205.21 | |
FHOAKXBXYSJBGX-MLHKIVSYNA-N | |
98766 |
C8H15NO5 | |
MFCD00037243 | |
boc-ser-oh, n-boc-l-serine, boc-l-serine, n-tert-butoxycarbonyl-l-serine, n-boc-serine, s-n-boc-serine, n-tert-butoxycarbonyl serine, s-2-tert-butoxycarbonyl amino-3-hydroxypropanoic acid, n-t-butoxycarbonyl-l-serine, boc-l-ser | |
CC(C)(C)OC(=O)N[C@@H](CO)C(O)=O |
RUO – Research Use Only