missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Serine benzyl ester, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 434840010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
BOC-L-Serine benzyl ester | |
Authentic | |
Glass bottle | |
1g | |
boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester | |
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1 | |
295.33 | |
295.33 |
59524-02-6 | |
97.5% min. (HPLC) | |
C15H21NO5 | |
-14° to -18° (20°C, 589nm) (c=1, ethanol) | |
BQADRZHPZVQGCW-LBPRGKRZSA-N | |
benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate | |
7018792 | |
98% |
Chemical Identifiers
59524-02-6 | |
295.33 | |
boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester | |
benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate |
C15H21NO5 | |
BQADRZHPZVQGCW-LBPRGKRZSA-N | |
7018792 | |
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1 |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only