missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-L-Proline, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC275651000

 View more versions of this product

Catalog No. AC275651000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

BOC-L-Proline
99+%
Authentic
22, 35
Glass bottle
CC(C)(C)OC(=O)N1CCCC1C(=O)O
215.249
215.25
Crystals or Crystalline Powder
0.5% max. (K.F.)
White
100g
15761-39-4
C10H17NO4
MFCD00037324
1-tert-butoxycarbonyl-l-proline, boc-l-pro, boc-l-pro-oh, boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, n-tert-butyloxycarbonyl-l-proline, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, tert-butoxycarbonyl-l-proline
ZQEBQGAAWMOMAI-ZETCQYMHSA-N
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
85083
− 60.50
≥99%
−60.5° (20°C 589nm,c=2,CH3COOH)
133.0°C to 136.0°C

Chemical Identifiers

15761-39-4
215.249
ZQEBQGAAWMOMAI-ZETCQYMHSA-N
85083
CC(C)(C)OC(=O)N1CCCC1C(=O)O
C10H17NO4
MFCD00037324
1-tert-butoxycarbonyl-l-proline, boc-l-pro, boc-l-pro-oh, boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, n-tert-butyloxycarbonyl-l-proline, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, tert-butoxycarbonyl-l-proline
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SDS Documents