missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Proline, 99+%, Thermo Scientific™
Chemical Identifiers
CAS | 15761-39-4 |
---|---|
Molecular Formula | C10H17NO4 |
Molecular Weight (g/mol) | 215.249 |
MDL Number | MFCD00037324 |
InChI Key | ZQEBQGAAWMOMAI-ZETCQYMHSA-N |
Synonym | boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro |
PubChem CID | 85083 |
IUPAC Name | (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
SMILES | CC(C)(C)OC(=O)N1CCCC1C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC275650250
|
Thermo Scientific Chemicals
275650250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC275651000
|
Thermo Scientific Chemicals
275651000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
15761-39-4 | |
215.249 | |
ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
85083 | |
CC(C)(C)OC(=O)N1CCCC1C(=O)O |
C10H17NO4 | |
MFCD00037324 | |
boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Specifications
15761-39-4 | |
C10H17NO4 | |
MFCD00037324 | |
Glass bottle | |
ZQEBQGAAWMOMAI-ZETCQYMHSA-N | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
215.249 | |
215.25 | |
0.5% max. (K.F.) | |
BOC-L-Proline |
99+% | |
Authentic | |
22, 35 | |
boc-l-proline, boc-pro-oh, n-boc-l-proline, n-tert-butoxycarbonyl-l-proline, boc-l-pro-oh, s-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butyloxycarbonyl-l-proline, tert-butoxycarbonyl-l-proline, 1-tert-butoxycarbonyl-l-proline, boc-l-pro | |
CC(C)(C)OC(=O)N1CCCC1C(=O)O | |
− 60.50 | |
85083 | |
≥99% | |
25g |
RUO â Research Use Only