missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Methionine, +99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 275630050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
47.0°C to 53.0°C | |
99+% | |
White | |
C10H19NO4S | |
CH3SCH2CH2CH(COOH)NHCOOC(CH3)3 | |
Glass Bottle | |
IMUSLIHRIYOHEV-ZETCQYMHSA-N | |
(2S)-2-{[(tert-butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid | |
249.33 | |
249.33 | |
0.5% max. (K.F.) | |
Powder |
BOC-L-Methionine | |
2488-15-5 | |
99+% | |
Authentic | |
MFCD00065586 | |
boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl | |
CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
− 23.00 | |
89857 | |
≥99% | |
5 g |
Chemical Identifiers
2488-15-5 | |
249.33 | |
IMUSLIHRIYOHEV-ZETCQYMHSA-N | |
89857 |
C10H19NO4S | |
MFCD00065586 | |
boc-l-methionine, boc-met-oh, n-boc-l-methionine, n-tert-butoxycarbonyl-l-methionine, tert-butoxycarbonyl-l-methionine, s-2-tert-butoxycarbonyl amino-4-methylthio butanoic acid, n-alpha-t-butyloxycarbonyl-l-methionine, methionine, n-1,1-dimethylethoxy carbonyl, l-methionine, n-1,1-dimethylethoxy carbonyl | |
CSCC[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Safety and Handling
EINECSNumber : 219-639-3
RUO – Research Use Only