missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Leucine monohydrate, 99%, Thermo Scientific™
Chemical Identifiers
CAS | 200936-87-4 |
---|---|
Molecular Formula | C11H21NO4·H2O |
Molecular Weight (g/mol) | 249.31 |
InChI Key | URQQEIOTRWJXBA-UHFFFAOYNA-N |
Synonym | boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine |
PubChem CID | 83170 |
ChEBI | CHEBI:28702 |
IUPAC Name | (2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
SMILES | O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC435490250
|
Thermo Scientific Chemicals
435490250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC435491000
|
Thermo Scientific Chemicals
435491000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
200936-87-4 | |
249.31 | |
boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine | |
CHEBI:28702 | |
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O |
C11H21NO4·H2O | |
URQQEIOTRWJXBA-UHFFFAOYNA-N | |
83170 | |
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
Specifications
200936-87-4 | |
98.5% min. (HPLC) | |
C11H21NO4·H2O | |
25g | |
-24.5 | |
URQQEIOTRWJXBA-UHFFFAOYNA-N | |
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
83170 | |
249.31 | |
BOC-L-Leucine monohydrate |
Authentic | |
Glass bottle | |
(CH3)2CHCH2CH(COOH)NHCO2C(CH3)3·H2O | |
−24.50° (25°C c=2,acetic acid) | |
boc-leu-oh, boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-boc-l-leucine, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, n-tert-butoxycarbonylleucine, boc-l-leu-oh, tert-butyloxycarbonyl-l-leucine | |
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O | |
249.31 | |
CHEBI:28702 | |
99% |
RUO â Research Use Only