missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Leucine monohydrate, 99%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC435490250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
BOC-L-Leucine monohydrate | |
White | |
98.5% min. (HPLC) | |
C11H21NO4·H2O | |
Glass bottle | |
25g | |
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O | |
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
249.31 | |
CHEBI:28702 | |
Powder | |
6 to 8% (KF) |
200936-87-4 | |
82°C to 90°C | |
Authentic | |
(CH3)2CHCH2CH(COOH)NHCO2C(CH3)3·H2O | |
boc-l-leu-oh, boc-l-leucine, boc-leu-oh, n-boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, tert-butyloxycarbonyl-l-leucine | |
URQQEIOTRWJXBA-UHFFFAOYNA-N | |
−24.50° (25°C c=2,acetic acid) | |
-24.5 | |
83170 | |
249.31 | |
99% |
Chemical Identifiers
200936-87-4 | |
249.31 | |
boc-l-leu-oh, boc-l-leucine, boc-leu-oh, n-boc-l-leucine, n-tert-butoxycarbonyl-l-leucine, n-tert-butoxycarbonylleucine, s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid, tert-butoxycarbonyl-l-leucine, tert-butoxycarbonylleucine, tert-butyloxycarbonyl-l-leucine | |
CHEBI:28702 | |
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O |
C11H21NO4·H2O | |
URQQEIOTRWJXBA-UHFFFAOYNA-N | |
83170 | |
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
RUO – Research Use Only