missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-L-Leucine monohydrate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435490050

 View more versions of this product

Catalog No. AC435490050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

BOC-L-Leucine monohydrate
Glass Bottle
-22.5° to -27.5° (20°C, 589nm) (c=2, glacial acetic acid)
White
5g
99%
(CH3)2CHCH2CH(COOH)nHCO2C(CH3)3·H2O
MDXGYYOJGPFFJL-QMMMGPOBSA-N
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
83170
249.31
≥98.5%
Authentic
−24.50° (25°C c=2,acetic acid)
6 to 8% (KF)
85°C to 90°C
13139-15-6
C11H21NO4
boc-l-leu-oh
CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C
231.292
CHEBI:28702
Powder

Chemical Identifiers

13139-15-6
231.292
boc-l-leu-oh
CHEBI:28702
CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C
C11H21NO4
MDXGYYOJGPFFJL-QMMMGPOBSA-N
83170
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SDS Documents