Please login to your online account to display your discounted pricing

BOC-L-Glutamic acid 5-benzylester dicyclohexylamine salt, 99.5+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  275560250

 View more versions of this product

Catalog No. AC275560250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.50%
Physical Form Amorphous Powder
Formula Weight 518.69
Quantity 25g
Color White
EINECS Number 237-008-0
Melting Point 140° to 143°C
Chemical Name or Material BOC-L-Glutamic acid 5-benzylester dicyclohexylamine salt
Packaging Glass bottle
Molecular Formula C17 H23 N O6 . C12 H23 N
Linear Formula C6H5CH2OCO(CH2)2CHCOOHNHCOOC(CH3)3·DCHA
Name Note 99.5+%
CAS 13574-84-0
Assay Percent Range 99.5+%
MDL Number MFCD00051281
Synonym N-(tert-Butoxycarbonyl)-L-glutamic acid 5-benzylester dicyclohexylamine salt
Specific Rotation -18.5° to -19.5° (21°C, 589nm) (c=2, CH3OH)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material BOC-L-Glutamic acid 5-benzylester dicyclohexylamine salt
Name Note 99.5+%
CAS 13574-84-0
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C17 H23 N O6 . C12 H23 N
Linear Formula C6H5CH2OCO(CH2)2CHCOOHNHCOOC(CH3)3·DCHA
Formula Weight 518.69
MDL Number MFCD00051281
Synonym N-(tert-Butoxycarbonyl)-L-glutamic acid 5-benzylester dicyclohexylamine salt