missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-L-Glutamic acid 5-benzylester dicyclohexylamine salt, 99.5+%, ACROS Organics™

Chemical Identifiers

CAS 13574-84-0
Molecular Formula C29H46N2O6
Molecular Weight (g/mol) 518.695
MDL Number MFCD00051281
InChI Key PLGYLOMCRIZGSY-ZOWNYOTGSA-N
Synonym 2s-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid; dicha, 5-benzyl n-tert-butoxycarbonyl-l-2-aminoglutarate, compound with n-dicyclohexylamine 1:1, boc-glu obzl-oh dicyclohexylammonium salt, boc-l-glutamic acid 5-benzyl ester dicyclohexylammonium salt, dicyclohexylamine s-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoate, n-cyclohexylcyclohexanamine; 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-oxo-5-phenylmethoxypentanoic acid
PubChem CID 2724759
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC275560050
SDS View Documents
Thermo Fisher Scientific
275560050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13574-84-0
518.695
PLGYLOMCRIZGSY-ZOWNYOTGSA-N
2724759
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
C29H46N2O6
MFCD00051281
2s-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid; dicha, 5-benzyl n-tert-butoxycarbonyl-l-2-aminoglutarate, compound with n-dicyclohexylamine 1:1, boc-glu obzl-oh dicyclohexylammonium salt, boc-l-glutamic acid 5-benzyl ester dicyclohexylammonium salt, dicyclohexylamine s-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoate, n-cyclohexylcyclohexanamine; 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-oxo-5-phenylmethoxypentanoic acid
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid

Specifications

Authentic
-18.5° to -19.5° (21°C, 589nm) (c=2, CH3OH)
140°C to 143°C
13574-84-0
C29H46N2O6
MFCD00051281
PLGYLOMCRIZGSY-ZOWNYOTGSA-N
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
2724759
Amorphous Powder
BOC-L-Glutamic acid 5-benzylester dicyclohexylamine salt, 99.5+%
Glass Bottle
White
5g
99.5+%
C6H5CH2OCO(CH2)2CHCOOHNHCOOC(CH3)3·DCHA
2s-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid; dicha, 5-benzyl n-tert-butoxycarbonyl-l-2-aminoglutarate, compound with n-dicyclohexylamine 1:1, boc-glu obzl-oh dicyclohexylammonium salt, boc-l-glutamic acid 5-benzyl ester dicyclohexylammonium salt, dicyclohexylamine s-5-benzyloxy-2-tert-butoxycarbonyl amino-5-oxopentanoate, n-cyclohexylcyclohexanamine; 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-oxo-5-phenylmethoxypentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(=O)O.C1CCC(CC1)NC2CCCCC2
518.695
518.69
≥99.50%
Safety and Handling

Safety and Handling

EINECSNumber : 237-008-0

SDS