Please login to your online account to display your discounted pricing

BOC-L-Aspartic acid 4-methyleester, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC426230010

 View more versions of this product

Catalog No. AC426230010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Assay ≥97.5% (Acid)
Formula Weight 247.25
Quantity 1g
Color Clear colorless to yellow
Chemical Name or Material BOC-L-Aspartic acid 4-methyleester
Assay Percent Range 98%
Infrared Spectrum Authentic
Molecular Formula C10H17NO6
CAS 59768-74-0
Synonym BOC-Asp(OMe)-OH
Specific Rotation −5° to −7° (20°C, 589nm) (c=2, MeOH)

Specifications

Chemical Name or Material BOC-L-Aspartic acid 4-methyleester
Assay ≥97.5% (Acid)
Assay Percent Range 98%
Molecular Formula C10H17NO6
Formula Weight 247.25
Synonym BOC-Asp(OMe)-OH