Please login to your online account to display your discounted pricing

BOC-L-Aspartic acid 4-methyleester, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material BOC-L-Aspartic acid 4-methyleester
Assay ≥97.5% (Acid)
Assay Percent Range 98%
Molecular Formula C10H17NO6
Formula Weight 247.25
Synonym BOC-Asp(OMe)-OH
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC426230050

  SDS
thermo fisher scientific
AC426230050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426230010

  SDS
thermo fisher scientific
AC426230010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 247.25
Assay ≥97.5% (Acid)
Quantity 5g
Color Clear colorless to yellow
Specific Rotation −5° to −7° (20°C, 589nm) (c=2, MeOH)
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C10H17NO6
CAS 59768-74-0
Chemical Name or Material BOC-L-Aspartic acid 4-methyleester
Synonym BOC-Asp(OMe)-OH
Infrared Spectrum Authentic