missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Aspartic acid 1-benzyl ester, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440820250
Specifications
BOC-L-Aspartic acid 1-benzyl ester | |
− 23.40 | |
White | |
25g | |
97.5% min. (HPLC) | |
1-benzyl n-boc-l-aspartate, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp-obzl, boc-l-asp h-obn, boc-l-aspartic acid 1-benzyl ester, commericial, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, pubchem12927 | |
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1 | |
323.34 | |
323.34 | |
98% |
Authentic | |
1% Max. | |
96.0°C to 99.0°C | |
30925-18-9 | |
C16H21NO6 | |
LDRWTKQWSXGSTM-LBPRGKRZSA-N | |
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid | |
7017887 | |
Powder |
Chemical Identifiers
30925-18-9 | |
323.34 | |
1-benzyl n-boc-l-aspartate, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp-obzl, boc-l-asp h-obn, boc-l-aspartic acid 1-benzyl ester, commericial, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, pubchem12927 | |
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid |
C16H21NO6 | |
LDRWTKQWSXGSTM-LBPRGKRZSA-N | |
7017887 | |
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse
GHS Signal Word: Warning