missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-L-Aspartic acid 1-benzyl ester, 98%, ACROS Organics™

Chemical Identifiers

CAS 30925-18-9
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.34
InChI Key LDRWTKQWSXGSTM-LBPRGKRZSA-N
Synonym 1-benzyl n-boc-l-aspartate, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp-obzl, boc-l-asp h-obn, boc-l-aspartic acid 1-benzyl ester, commericial, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, pubchem12927
PubChem CID 7017887
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC440820050
SDS View Documents
Thermo Fisher Scientific
AC440820050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC440820250
SDS View Documents
Thermo Fisher Scientific
AC440820250
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

30925-18-9
323.34
1-benzyl n-boc-l-aspartate, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp-obzl, boc-l-asp h-obn, boc-l-aspartic acid 1-benzyl ester, commericial, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, pubchem12927
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
C16H21NO6
LDRWTKQWSXGSTM-LBPRGKRZSA-N
7017887
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Authentic
1% Max.
96.0°C to 99.0°C
30925-18-9
C16H21NO6
LDRWTKQWSXGSTM-LBPRGKRZSA-N
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
7017887
Powder
BOC-L-Aspartic acid 1-benzyl ester
− 23.40
White
5g
97.5% min. (HPLC)
1-benzyl n-boc-l-aspartate, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-asp-obzl, boc-l-asp h-obn, boc-l-aspartic acid 1-benzyl ester, commericial, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, pubchem12927
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1
323.34
323.34
98%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse

GHS Signal Word: Warning