missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-L-Alanine, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 275490100
Specifications
BOC-L-Alanine | |
99+% | |
(CH3)3COC(O)NHC(CH3)COOH | |
MFCD00037225 | |
boc-ala, boc-ala-oh, boc-l-ala-oh, boc-l-alanine, n-alpha-t-boc-l-alanine, n-boc-l-alanine, n-t-boc-l-alanine, n-tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, tert-butoxycarbonyl-l-alanine | |
QVHJQCGUWFKTSE-YFKPBYRVSA-N | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
85082 | |
− 26.00 | |
≥99% | |
81.0°C to 84.0°C |
15761-38-3 | |
C8H15NO4 | |
Authentic | |
04, 2507 | |
Glass bottle | |
CC(C(=O)O)NC(=O)OC(C)(C)C | |
189.211 | |
189.21 | |
Granular Powder | |
White | |
10g |
Chemical Identifiers
15761-38-3 | |
189.211 | |
QVHJQCGUWFKTSE-YFKPBYRVSA-N | |
85082 | |
CC(C(=O)O)NC(=O)OC(C)(C)C |
C8H15NO4 | |
MFCD00037225 | |
boc-ala, boc-ala-oh, boc-l-ala-oh, boc-l-alanine, n-alpha-t-boc-l-alanine, n-boc-l-alanine, n-t-boc-l-alanine, n-tert-butoxycarbonyl-l-alanine, tert-butoxycarbonyl alanine, tert-butoxycarbonyl-l-alanine | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |