missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-Glycine N-hydroxysuccinimide ester, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC436700050

 View more versions of this product

Catalog No. AC436700050



Specifications

Specifications

(0.3g in 6.0mL THF) Clear
Authentic
158.4°C to 161.9°C
3392-07-2
C11H16N2O6
LJCWRJYVPJJTMB-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
76926
Powder
BOC-Glycine N-hydroxysuccinimide ester
White
5g
99%
boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, bocglyosu, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate
CC(C)(C)OC(=O)NCC(=O)ON1C(=O)CCC1=O
272.26
272.26
99%

Chemical Identifiers

3392-07-2
272.26
boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, bocglyosu, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate
(2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
C11H16N2O6
LJCWRJYVPJJTMB-UHFFFAOYSA-N
76926
CC(C)(C)OC(=O)NCC(=O)ON1C(=O)CCC1=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents