missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-Glycine N-hydroxysuccinimide ester, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436700010

 View more versions of this product

Catalog No. AC436700010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(0.3g in 6.0mL THF) Clear
(0.3g in 6.0 ml THF) Clear
Glass Bottle
158.4°C to 161.9°C
3392-07-2
C11H16N2O6
LJCWRJYVPJJTMB-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
76926
Powder
BOC-Glycine N-hydroxysuccinimide ester
Authentic
White
1g
97.5% min. (GC)
2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate, boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, bocglyosu, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate
CC(C)(C)OC(=O)NCC(=O)ON1C(=O)CCC1=O
272.26
272.26
99%

Chemical Identifiers

3392-07-2
272.26
2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate, boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, bocglyosu, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate
(2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
C11H16N2O6
LJCWRJYVPJJTMB-UHFFFAOYSA-N
76926
CC(C)(C)OC(=O)NCC(=O)ON1C(=O)CCC1=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-230-2

SDS Documents