Please login to your online account to display your discounted pricing

BOC-Glycine N-hydroxysuccinimide ester, 99%, ACROS Organics™

$137.51 - $137.51

Chemical Identifiers

Chemical Name or Material BOC-Glycine N-hydroxysuccinimide ester
Assay ≥97.5%
Assay Percent Range 99%
Molecular Formula C11H16N2O6
Formula Weight 272.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC436700050

 
thermo fisher scientific
AC436700050
5g Each for $137.51

AC436700010

 
thermo fisher scientific
AC436700010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 272.26
Assay ≥97.5%
Quantity 5g
Color White
Melting Point 152° to 167°C
Clarity (0.3g in 6.0mL THF) Clear
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material BOC-Glycine N-hydroxysuccinimide ester
Molecular Formula C11H16N2O6
Assay Percent Range 99%
CAS 3392-07-2
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.