Please login to your online account to display your discounted pricing

BOC-Glycine N-hydroxysuccinimide ester, 99%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  436700250

 View more versions of this product

Catalog No. AC436700250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Clarity (0.3g in 6.0mL THF) Clear
Infrared Spectrum Authentic
Melting Point 152° to 167°C
Quantity 25g
Color White
Chemical Name or Material BOC-Glycine N-hydroxysuccinimide Ester
CAS 3392-07-2
Physical Form White powder
Assay 97.5% min.
Assay Percent Range 99%
Molecular Formula C11H16N2O6
Formula Weight 272.26g/mol
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material BOC-Glycine N-hydroxysuccinimide Ester
Assay 97.5% min.
Assay Percent Range 99%
Molecular Formula C11H16N2O6
Formula Weight 272.26g/mol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.