missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BOC-DL-Valine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 425780500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
BOC-DL-Valine | |
97.5% min. (HPLC) | |
C10H19NO4 | |
MFCD00069973 | |
boc-dl-valine, boc-dl-val-oh, n-boc-dl-valine, 2-tert-butoxycarbonyl amino-3-methylbutanoic acid, dl-n-tert-butoxycarbonyl valine, n-tert-butoxycarbonyl valine, 2-tert-butoxy carbonyl amino-3-methylbutanoic acid, n-tert-butoxycarbonyl-dl-valine, n-alpha-t-boc-l-valine | |
CC(C)C(C(=O)O)NC(=O)OC(C)(C)C | |
217.26 | |
217.26 |
54895-12-4 | |
Glass bottle | |
(CH3)2CHCH(COOH)NHCOOC(CH3)3 | |
50g | |
SZXBQTSZISFIAO-UHFFFAOYSA-N | |
3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
270655 | |
98% |
Chemical Identifiers
54895-12-4 | |
217.26 | |
SZXBQTSZISFIAO-UHFFFAOYSA-N | |
270655 | |
CC(C)C(C(=O)O)NC(=O)OC(C)(C)C |
C10H19NO4 | |
MFCD00069973 | |
boc-dl-valine, boc-dl-val-oh, n-boc-dl-valine, 2-tert-butoxycarbonyl amino-3-methylbutanoic acid, dl-n-tert-butoxycarbonyl valine, n-tert-butoxycarbonyl valine, 2-tert-butoxy carbonyl amino-3-methylbutanoic acid, n-tert-butoxycarbonyl-dl-valine, n-alpha-t-boc-l-valine | |
3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
RUO â Research Use Only