Please login to your online account to display your discounted pricing

BOC-D-Methionine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material BOC-D-Methionine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H19NO4S
Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
Formula Weight 249.32
MDL Number MFCD00038256
Synonym N-(tert-Butoxycarbonyl)-D-methionine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336410250

  SDS
thermo fisher scientific
AC336410250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC336410010

  SDS
fisher scientific acros
336410010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 249.32
Quantity 25g
Color White to off-white
Specific Rotation Condition +23.00 (20.00°C c=1.3,methanol)
Melting Point 49° to 55°C
Assay Percent Range 98%
Water 0.5% max. (K.F.)
Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
Chemical Name or Material BOC-D-Methionine
Packaging Glass bottle
Specific Rotation +21.5° to +24.5° (20°C, 589nm) (c=1, CH3OH)
Molecular Formula C10H19NO4S
MDL Number MFCD00038256
CAS 5241-66-7
Synonym N-(tert-Butoxycarbonyl)-D-methionine
Infrared Spectrum Authentic