Please login to your online account to display your discounted pricing

BOC-D-Methionine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  336410010

 View more versions of this product

Catalog No. AC336410010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 249.32
Physical Form Powder
Quantity 1g
Color White to off-white
Melting Point 49° to 55°C
Chemical Name or Material BOC-D-Methionine
Packaging Glass bottle
Assay Percent Range 98%
Water 0.5% max. (K.F.)
Specific Rotation +21.5° to +24.5° (20°C, 589nm) (c=1, CH3OH)
Molecular Formula C10H19NO4S
Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
MDL Number MFCD00038256
CAS 5241-66-7
Synonym N-(tert-Butoxycarbonyl)-D-methionine
Infrared Spectrum Authentic

Specifications

Chemical Name or Material BOC-D-Methionine
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H19NO4S
Linear Formula CH3SCH2CH2CH(COOH)NHCOOC(CH3)3
Formula Weight 249.32
MDL Number MFCD00038256
Synonym N-(tert-Butoxycarbonyl)-D-methionine