missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BOC-D-α-cyclohexylglycine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC399360050

 View more versions of this product

Catalog No. AC399360050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

257.33
MFCD00133629
70491-05-3
98%
BOC-D-Chg-OH
5g
C13H23NO4
BOC-D-α-Cyclohexylglycine
Glass bottle

Chemical Identifiers

98%
257.33
BOC-D-Chg-OH
C13H23NO4
MFCD00133629
BOC-D-α-Cyclohexylglycine