Please login to your online account to display your discounted pricing

BOC-D-α-cyclohexylglycine, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC399360010

 View more versions of this product

Catalog No. AC399360010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 257.33
Quantity 1g
Chemical Name or Material BOC-D-α-Cyclohexylglycine
Assay Percent Range 98%
Molecular Formula C13H23NO4
MDL Number MFCD00133629
CAS 70491-05-3
Packaging Glass bottle
Synonym BOC-D-Chg-OH

Specifications

Chemical Name or Material BOC-D-α-Cyclohexylglycine
Assay Percent Range 98%
Molecular Formula C13H23NO4
Formula Weight 257.33
MDL Number MFCD00133629
Synonym BOC-D-Chg-OH