missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(BOC-aminooxy)acetic acid, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC429690010

 View more versions of this product

Catalog No. AC429690010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(BOC-Aminooxy)acetic acid
111.5°C to 112.0°C
42989-85-5
100.0
C7H13NO5
2-2-methylpropan-2-yl oxycarbonylamino oxyacetic acid
CC(C)(C)OC(=O)NOCC(=O)O
191.19
191.19
Glass bottle
1g
97.5
98%
MFCD01632027
QBXODCKYUZNZCY-UHFFFAOYSA-N
2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetic acid
2755974
98%

Chemical Identifiers

42989-85-5
191.19
QBXODCKYUZNZCY-UHFFFAOYSA-N
2755974
CC(C)(C)OC(=O)NOCC(=O)O
C7H13NO5
MFCD01632027
2-2-methylpropan-2-yl oxycarbonylamino oxyacetic acid
2-[(2-methylpropan-2-yl)oxycarbonylamino]oxyacetic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents